Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chloro-2-nitrophenol, (max. 10% H₂O) ≥98%

4-Chloro-2-nitrophenol, (max. 10% H₂O) ≥98%

Glass bottle for solids, 25 g
Catalog # A18150.14
Irritant
4-Chloro-2-nitrophenol, (max. 10% H₂O) ≥98%
Catalog # A18150.14
CAS Number:  89-64-5
4-Chloro-2-nitrophenol, (max. 10% H₂O) ≥98%
4-Chloro-2-nitrophenol, (max. 10% H₂O) ≥98%

Glass bottle for solids, 25 g

CAS Number:  89-64-5
Price
35,90 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 89-64-5 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Melting Point: 85-88° Beilstein Reference: 2048031

  • CAS Number:89-64-5
  • Formula:C₆H₄ClNO₃
  • MDL Number:MFCD00007113
  • Molecular Weight:173.56 g/mol
  • Melting Pt:85…88 °C
  • Storage Temperature:Ambient
  • EINECS:201-927-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications