Order Entry
Export
ContactUsLinkComponent
 
1,3-Dichloro-4-nitrobenzene
Catalog # EHERC12432400
Supplier:  EHRENSTORFER
CAS Number:  
1,3-Dichloro-4-nitrobenzene
Catalog # EHERC12432400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :192 g/mol
Boiling Pt:258 °C (1013 hPa)
Melting Pt:28…31 °C
Density:1.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-248-3
UN:2811
ADR:6.1,III
CAS Number:611-06-3
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD00007071