5-Chloro-2-nitroaniline
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:1635-61-6
:
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD00007776
:172.57 g/mol
Boiling Pt:323 °C (1013 hPa)
Melting Pt:123…126 °C
EINECS:216-661-5
UN:2237
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient