


2,5-Dichloro-4-nitroaniline
Catalog # MOLE10250092-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.02 g/mol
Storage Temperature:Ambient
EINECS:229-591-5
UN:2237
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:6627-34-5
Formula:C₆H₄Cl₂N₂O₂
MDL Number:MFCD00025157