


2,5-Dichloro-4-nitroaniline
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:6627-34-5
:
Formula:C₆H₄Cl₂N₂O₂
MDL Number:MFCD00025157
:207.02 g/mol
Storage Temperature:Ambient
EINECS:229-591-5
UN:2237
ADR:6.1,III
:GHS06
:GHS08
:GHS09