Order Entry
Export
ContactUsLinkComponent
3,4-Dichloroaniline
Toxic
Corrosive
Environment
3,4-Dichloroaniline
Catalog # MOLEM61344960
Supplier:  Molekula
CAS Number:  
Toxic
Corrosive
Environment
3,4-Dichloroaniline
Catalog # MOLEM61344960
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :162.02 g/mol
Merck Index:13,03079
Boiling Pt:272 °C (1013 hPa)
Melting Pt:70…73 °C
Density:1.567 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-448-4
UN:3442
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS09
CAS Number:95-76-1
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007768