Order Entry
Export
ContactUsLinkComponent
 
 
 
 
2,6-Dichloroaniline 98%
 
undefined
 
 
 
2,6-Dichloroaniline 98%

 

  • Appearance:
    White to brown Crystalline solid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    35 to 39 °C
  • GC:
    ≥97.5 %
  • BSC Certification:
    Passes