Order Entry
Export
ContactUsLinkComponent
2,5-Dichloroaniline
Toxic
Health
Environment
2,5-Dichloroaniline
Catalog # MOLE32619825-1KG
Supplier:  Molekula
CAS Number:  
Toxic
Health
Environment
2,5-Dichloroaniline
Catalog # MOLE32619825-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :162.02 g/mol
Boiling Pt:251 °C (1013 hPa)
Melting Pt:46…50 °C
Density:1.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-455-2
UN:3442
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:95-82-9
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007667