Order Entry
Export
ContactUsLinkComponent
 
 
 
 
2,4-Dichloroaniline 98%
 
undefined
 
 
 
2,4-Dichloroaniline 98%

 

  • Appearance:
    Beige to light grey to light brown Powder or crystals or flakes and/or chunks (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    59 to 63 °C
  • GC:
    ≥97.5 %