Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,6-Trichloroaniline
2,4,6-Trichloroaniline
2,4,6-Trichloroaniline
2,4,6-Trichloroaniline
EHERC17700600
:
EHRENSTORFER
:
2,4,6-Trichloroaniline
EHERC17700600
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
196.46 g/mol
:
262 °C (1013 hPa)
:
74 °C
:
Ambient
:
211-219-8
:
2018
:
6.1,II
:
GHS06
:
GHS08
:
GHS09
:
634-93-5
:
C₆H₄Cl₃N
:
MFCD00007663