Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloro-5-iodobenzene
1,3-Dichloro-5-iodobenzene
1,3-Dichloro-5-iodobenzene
1,3-Dichloro-5-iodobenzene
Supplier:
Thermo Scientific
Certificates
Expand image
1,3-Dichloro-5-iodobenzene
Supplier:
Thermo Scientific
About this item
CAS Number:
3032-81-3
:
Formula:
C₆H₃Cl₂I
MDL Number:
MFCD00001047
:
272.9 g/mol
Boiling Pt:
127 °C (20 mmHg)
Melting Pt:
57 °C
EINECS:
221-216-3