Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloro-4-iodobenzene
1,3-Dichloro-4-iodobenzene
1,3-Dichloro-4-iodobenzene
Irritant
1,3-Dichloro-4-iodobenzene
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
1,3-Dichloro-4-iodobenzene
Supplier:
Thermo Scientific
About this item
CAS Number:
29898-32-6
:
Formula:
C₆H₃Cl₂I
MDL Number:
MFCD00001035
:
272.9 g/mol
Boiling Pt:
262 °C (1013 hPa)
Density:
2,01 g/cm³ (20 °C)
EINECS:
249-938-4
:
GHS07