Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chloro-1,2-difluorobenzene ≥98%

4-Chloro-1,2-difluorobenzene ≥98%

Flammable
Irritant
4-Chloro-1,2-difluorobenzene ≥98%
Certificates
4-Chloro-1,2-difluorobenzene ≥98%
4-Chloro-1,2-difluorobenzene ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 696-02-6 Molecular Formula: C6H3ClF2 Formula Weight: 148.54 Flash Point: 32°(89°F) Refractive Index: 1.4745 Odour: Aromatic Boiling Point: 128-130° Beilstein Reference: 2081081 UN No.: 1993 MDL No.: MFCD00042572

Synonyms:  3,4-Difluorochlorobenzene
  • CAS Number:696-02-6
  • Formula:C₆H₃ClF₂
  • MDL Number:MFCD00042572
  • Molecular Weight:148.54 g/mol
  • Boiling Pt:128…130 °C
  • Density:1.350
  • Flash Pt:32 °C (89 °F)
  • Storage Temperature:Ambient
  • ADR:3,III
Product Family Options

Product Information