
3,5-Dichlorobenzoic acid
Catalog # EHERC12403100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:191.01 g/mol
Boiling Pt:309 °C (1013 hPa)
Melting Pt:183…185 °C
EINECS:200-092-4
:GHS07
Storage Temperature:Ambient
CAS Number:51-36-5
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002494