
3,5-Dichlorobenzoic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-36-5
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002494
:191.01 g/mol
Boiling Pt:309 °C (1013 hPa)
Melting Pt:185…187 °C
Storage Temperature:Ambient
EINECS:200-092-4
:GHS07