
2,6-Dichlorobenzoic acid 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-30-6
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002418
:191.01 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:141…145 °C
Storage Temperature:Ambient
EINECS:200-025-9
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:141 to 145 °C
- HPLC:≥98.0 %
- Titration with NaOH:≥98.0 %