
2,5-Dichlorobenzoic acid
Supplier: CHEMPUR
Certificates
About this item
CAS Number:50-79-3
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002416
:191.01 g/mol
Boiling Pt:301 °C (1013 hPa)
Melting Pt:151…154 °C
Storage Temperature:Ambient
EINECS:200-065-7
:GHS07