
2,4-Dichlorobenzoic acid
Catalog # EHERC12400000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:191.01 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:158…160 °C
EINECS:200-067-8
:GHS07
Storage Temperature:Ambient
CAS Number:50-84-0
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002414