

2,5-Dichlorobenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21663-61-6
:
Formula:C₇H₃Cl₂N
MDL Number:MFCD00019741
:172.01 g/mol
Boiling Pt:269 °C (1013 hPa)
Melting Pt:128…131 °C
Storage Temperature:Ambient
EINECS:244-518-7
UN:3439
ADR:6.1,III
:GHS06
:GHS07