Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorobenzonitrile
2,3-Dichlorobenzonitrile
2,3-Dichlorobenzonitrile
2,3-Dichlorobenzonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
2,3-Dichlorobenzonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
6574-97-6
:
Formula:
C₇H₃Cl₂N
MDL Number:
MFCD00016372
:
172.01 g/mol
Boiling Pt:
70 °C (0.5 mmHg)
Melting Pt:
59 °C
UN:
3439
ADR:
6.1,III