0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Chloro-3-nitrobenzaldehyde
4-Chloro-3-nitrobenzaldehyde ≥97%
4-Chloro-3-nitrobenzaldehyde ≥97%
:
Thermo Fisher Scientific
4-Chloro-3-nitrobenzaldehyde ≥97%
:
Thermo Fisher Scientific
:
16588-34-4
:
:
C₇H₄ClNO₃
:
MFCD00007078
:
185.57 g/mol
:
61…65 °C
:
240-645-7
:
GHS07
:
Ambient
msds_ALFAA13549
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years