4-Chloro-3-nitrobenzaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16588-34-4
:
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007078
:185.57 g/mol
Melting Pt:61…63 °C
EINECS:240-645-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to light yellow to light green Powder
- Infrared spectrum:Conforms
- Melting point:60 to 65 °C
- GC:≥96.0 %
- Titration with HClO4:98.5 to 101.5 %