
3,5-Dichlorobenzaldehyde
Catalog # MAYBSEW00693.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.01 g/mol
Boiling Pt:215…222 °C (7 mmHg)
Melting Pt:63 °C
EINECS:233-499-0
:GHS07
Storage Temperature:Ambient
CAS Number:10203-08-4
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003352