
2,6-Dichlorobenzaldehyde
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:83-38-5
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003307
:175.01 g/mol
Boiling Pt:239 °C (1013 hPa)
Melting Pt:69…70 °C
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:201-472-2
:GHS07