2,3-Dichlorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
- CAS Number:6334-18-5
- Formula:C₇H₄Cl₂O
- MDL Number:MFCD00010127
- Molecular Weight:175.01 g/mol
- Boiling Pt:143…145 °C (30 mmHg)
- Melting Pt:61…65 °C
- Density:1.33
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:228-711-3
- ADR:8,III
2 Options Available Below
Product Details & Documents
- CAS Number:6334-18-5
- Formula:C₇H₄Cl₂O
- MDL Number:MFCD00010127
- Molecular Weight:175.01 g/mol
- Boiling Pt:143…145 °C (30 mmHg)
- Melting Pt:61…65 °C
- Density:1.33
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:228-711-3
- ADR:8,III



