Hexachlorobenzene
Glass bottle, 1,2 mL
Catalog # PROOCILCLM-351-1.2
Supplier: LGC Standards PROMOCHEM
Hexachlorobenzene
Catalog # PROOCILCLM-351-1.2
Supplier: LGC Standards PROMOCHEM
CAS Number: 118-74-1
Jump to:
Product Details & Documents
- CAS Number:118-74-1
- Formula:C₆Cl₆
- Molecular Weight:284.78 g/mol
- Merck Index:12,04714
- Boiling Pt:332 °C (1013 hPa)
- Melting Pt:2.045 °C
- Density:2.045 g/cm³ (20 °C)
- Flash Pt:242 °C
- Storage Temperature:Ambient
- EINECS:204-273-9
- ADR:6.1,III
- :Density: 2.045 g/cm³ (20 °C)
- UN:2729
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL



