1,3-Dichlorobenzene, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:541-73-1
- Formula:C₆H₄Cl₂
- MDL Number:MFCD00000573
- Molecular Weight:147 g/mol
- Merck Index:13,03080
- Boiling Pt:173 °C (1013 hPa)
- Melting Pt:–25 °C
- Density:1.2884 g/cm³ (20 °C)
- Flash Pt:65 °C
- Storage Temperature:Ambient
- EINECS:208-792-1
- UN:3082
- ADR:9,III
2 Options Available Below
Product Details & Documents
- CAS Number:541-73-1
- Formula:C₆H₄Cl₂
- MDL Number:MFCD00000573
- Molecular Weight:147 g/mol
- Merck Index:13,03080
- Boiling Pt:173 °C (1013 hPa)
- Melting Pt:–25 °C
- Density:1.2884 g/cm³ (20 °C)
- Flash Pt:65 °C
- Storage Temperature:Ambient
- EINECS:208-792-1
- UN:3082
- ADR:9,III



