

1,3-Dichlorobenzene
Catalog # EHERC12371000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Merck Index:13,03080
Boiling Pt:173 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.2884 g/cm³ (20 °C)
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:208-792-1
UN:3082
ADR:9,III
:147 g/mol
:GHS07
:GHS09
CAS Number:541-73-1
Formula:C₆H₄Cl₂
MDL Number:MFCD00000573