Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dichlorobenzene 98%

1,3-Dichlorobenzene 98%

Irritant
Environment
1,3-Dichlorobenzene 98%
Certificates
1,3-Dichlorobenzene 98%

4 Options Available Below

Product Details & Documents
  • CAS Number:541-73-1
  • Formula:C₆H₄Cl₂
  • MDL Number:MFCD00000573
  • Molecular Weight:147 g/mol
  • Merck Index:13,03080
  • Boiling Pt:173 °C (1013 hPa)
  • Melting Pt:–24 °C
  • Density:1.28 g/cm³
  • Flash Pt:67 °C
  • Storage Temperature:Ambient
  • EINECS:208-792-1
  • UN:3082
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5450 to 1.5470 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 21,10 €
    21,10 €
    1 * 25 mL
       21,10 €

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 60,70 €
    60,70 €
    1 * 250 mL
       60,70 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 97,10 €
    97,10 €
    1 * 500 mL
       97,10 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 161,00 €
    161,00 €
    1 * 1 L
       161,00 €