

1,2-Dichlorobenzene
Catalog # EHERC12370000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:147 g/mol
Merck Index:13,03081
Boiling Pt:180.5 °C (1013 hPa)
Melting Pt:–17 °C
Density:1.309 g/cm³ (20 °C)
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:202-425-9
UN:1591
ADR:6.1,III
:GHS07
:GHS09
CAS Number:95-50-1
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535