Gallic acid
Catalog # MOLEM41415749
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:170.12 g/mol
Merck Index:13,04366
Boiling Pt:501 °C (1013 hPa)
Melting Pt:251…252 °C
Density:1.694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-749-9
CAS Number:149-91-7
Formula:C₇H₆O₅
MDL Number:MFCD00002510