Order Entry
Export
ContactUsLinkComponent
3,4-Dihydroxybenzoic acid
3,4-Dihydroxybenzoic acid
Catalog # MOLE13192758-100G
Supplier:  Molekula
CAS Number:  
3,4-Dihydroxybenzoic acid
Catalog # MOLE13192758-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :154.12 g/mol
Merck Index:12,08080
Boiling Pt:125…135 °C (0.001 mmHg)
Melting Pt:202…204 °C
Density:1.5415 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-760-0
CAS Number:99-50-3
Formula:C₇H₆O₄
MDL Number:MFCD00002509