2,6-Dihydroxybenzoic acid
Catalog # MOLEM35122455
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:154.12 g/mol
Boiling Pt:339 °C (1013 hPa)
Melting Pt:152…158 °C
Density:1,58 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-134-8
CAS Number:303-07-1
Formula:C₇H₆O₄
MDL Number:MFCD00002462