2,5-Dihydroxybenzoic acid
Catalog # MOLEM22620684
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:154.12 g/mol
Merck Index:13,04409
Boiling Pt:339 °C (1013 hPa)
Melting Pt:202…206 °C
Density:1.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-718-5
CAS Number:490-79-9
Formula:C₇H₆O₄
MDL Number:MFCD00002460