Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dihydroxybenzoic acid

2,5-Dihydroxybenzoic acid

Glass bottle for solids, 100 g
Catalog # MOLE42641018-100G
Supplier:   Molekula
2,5-Dihydroxybenzoic acid
Catalog # MOLE42641018-100G
Supplier:  Molekula
CAS Number:  490-79-9
2,5-Dihydroxybenzoic acid

Glass bottle for solids, 100 g

CAS Number:  490-79-9
Price
125,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2,5-Dihydroxybenzoic Acid, DHB, Hydroquinonecarboxylic acid, Gentisic acid, 2,5-DHBA
  • CAS Number:490-79-9
  • Formula:C₇H₆O₄
  • MDL Number:MFCD00002460
  • Molecular Weight:154.12 g/mol
  • Merck Index:13,04409
  • Boiling Pt:339 °C (1013 hPa)
  • Melting Pt:202…206 °C
  • Density:1.55 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-718-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications