
3,5-Dimethoxybenzoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1132-21-4
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002502
:182.18 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:182…185 °C
Density:1,423 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-473-8
:GHS07
Specification Test Results
- Appearance:White to pink-beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:182 to 185 °C
- HPLC:≥98.5 %
- Titration with NaOH:≥98.5 %