Order Entry
Export
ContactUsLinkComponent
3,4-Dimethoxybenzoic acid
3,4-Dimethoxybenzoic acid
Catalog # MOLE36580840-100G
Supplier:  Molekula
CAS Number:  
3,4-Dimethoxybenzoic acid
Catalog # MOLE36580840-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :182.18 g/mol
Merck Index:12,10086
Boiling Pt:303 °C (1013 hPa)
Melting Pt:117…119 °C
Density:1.436 g/cm³ (–100 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:202-215-7
CAS Number:93-07-2
Formula:C₉H₁₀O₄
MDL Number:MFCD00002500