
3,4-Dimethoxybenzoic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:93-07-2
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002500
:182.18 g/mol
Merck Index:12,10086
Boiling Pt:303 °C (1013 hPa)
Melting Pt:179…182 °C
Density:1,436 g/cm³ (-100 °C)
Flash Pt:>100 °C
EINECS:202-215-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:180 to 184 °C
- HPLC:≥99.0 %
- Color scale:≤10 APHA