Order Entry
Export
ContactUsLinkComponent
 
Irritant
3,4-Dinitrobenzoic acid 99%
Catalog # ACRO408700050
CAS Number:  
undefined
Irritant
3,4-Dinitrobenzoic acid 99%
Catalog # ACRO408700050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    163 to 166 °C
  • HPLC:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %

Specification Test Results

About this item

 :
 :212.12 g/mol
Merck Index:13,03303
Melting Pt:163…166 °C
Density:1,674 g/cm³ (20 °C)
EINECS:208-433-9
 :GHS07
CAS Number:528-45-0
Formula:C₇H₄N₂O₆
MDL Number:MFCD00007255