Order Entry
Export
ContactUsLinkComponent
2,4-Dinitrobenzoic acid
Irritant
2,4-Dinitrobenzoic acid
Catalog # EHERC12783400
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
2,4-Dinitrobenzoic acid
Catalog # EHERC12783400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :212.12 g/mol
Melting Pt:179…183 °C
Density:1,672 g/cm³ (20 °C)
EINECS:210-219-5
 :GHS07
Storage Temperature:Ambient
CAS Number:610-30-0
Formula:C₇H₄N₂O₆
MDL Number:MFCD00017006