Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitro-o-anisic acid
4-Nitro-o-anisic acid ≥98%
4-Nitro-o-anisic acid ≥98%
Irritant
4-Nitro-o-anisic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
4-Nitro-o-anisic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
2597-56-0
:
Formula:
C₈H₇NO₅
MDL Number:
MFCD00216573
:
197.15 g/mol
Melting Pt:
145…148 °C
EINECS:
219-998-6
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB25523
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation