Order Entry
Export
ContactUsLinkComponent
3,5-Diaminobenzoic acid
3,5-Diaminobenzoic acid
Catalog # MOLE15796862-100G
Supplier:  Molekula
CAS Number:  
3,5-Diaminobenzoic acid
Catalog # MOLE15796862-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.15 g/mol
Merck Index:13,02997
Boiling Pt:546 °C (1013 hPa)
Melting Pt:239…242 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-621-0
CAS Number:535-87-5
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007807