
3,4-Diaminobenzoic acid
Catalog # MOLE28128705-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:152.15 g/mol
Boiling Pt:354 °C (1013 hPa)
Boiling Pt:354 °C (1013 hPa)
Melting Pt:211 °C
Density:1.544 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-577-2
:GHS07
CAS Number:619-05-6
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007726