4,5-Dimethoxyanthranilic acid
Supplier: CHEMPUR
Certificates
About this item
CAS Number:5653-40-7
:
Formula:C₉H₁₁NO₄
MDL Number:MFCD00011671
:197.19 g/mol
Boiling Pt:359 °C (1013 hPa)
Melting Pt:155…160 °C
Storage Temperature:Ambient
EINECS:227-095-3
:GHS07