Anthranilic acid
Glass bottle for solids, 1 kg
Catalog # MOLE15557294-1KG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Aminobenzoic acid
- CAS Number:118-92-3
- Formula:C₇H₇NO₂
- MDL Number:MFCD00007712
- Molecular Weight:137.14 g/mol
- Merck Index:13,00421
- Boiling Pt:312 °C (1013 hPa)
- Melting Pt:144 °C
- Density:1.362 g/cm³ (20 °C)
- Flash Pt:150 °C
- :Density: 1.362 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-287-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



