Anthranilic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Aminobenzoic acid
CAS Number:118-92-3
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007712
Boiling Pt:312
Melting Pt:144
Density:1.362 g/cm³ (20 °C)
Molecular Weight:137.14 g/mol
Flash Pt:150
Merck Index:13,00421
:M-Clarity™ quality level = MQ200
EINECS:204-287-5
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:5.7 g/l (25 °C)
- Melting Point:146 - 148 °C
- Molar Mass:137.14 g/mol
- Bulk Density:500 kg/m3
- Vapor Pressure:0.001 hPa (52.6 °C)
- Flash Point:150 °C
- Density:1.41 g/cm3 (20 °C)
- pH:4 (100 g/l, H2O, 20 °C)
- Ignition Point:530 °C