Organic reference standard, Benzoic acid 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA10537500AC
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:65-85-0
- Description:Benzoic acid 100 µg/ml in acetone
- Formula:C₇H₆O₂
- Description:Benzoic acid