2,6-Dimethoxybenzonitrile 97%
:16932-49-3
:
:C₉H₉NO₂
:MFCD00001788
:163.18 g/mol
:310 °C (1013 hPa)
:119…123 °C
:Ambient
:241-000-2
:2811
:6.1,III
:GHS07
- Appearance:Light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:119 to 123 °C
- GC:≥96.0 %
- HPLC:≥97.5 %