Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethoxybenzonitrile
2,4-Dimethoxybenzonitrile ≥99%
2,4-Dimethoxybenzonitrile ≥99%
:
Thermo Fisher Scientific
2,4-Dimethoxybenzonitrile ≥99%
:
Thermo Fisher Scientific
:
4107-65-7
:
:
C₉H₉NO₂
:
MFCD00001786
:
163.18 g/mol
:
283 °C (1013 hPa)
:
92…95 °C
:
Ambient
:
223-886-2
:
3439
:
6.1,III
:
GHS06
:
GHS07
msds_ALFAB24621
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years