Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethoxybenzonitrile
2,3-Dimethoxybenzonitrile
2,3-Dimethoxybenzonitrile
2,3-Dimethoxybenzonitrile
Supplier:
CHEMPUR
Certificates
Expand image
2,3-Dimethoxybenzonitrile
Supplier:
CHEMPUR
About this item
CAS Number:
5653-62-3
:
Formula:
C₉H₉NO₂
MDL Number:
MFCD00001784
Boiling Pt:
293 °C (1013 hPa)
Melting Pt:
47 °C
EINECS:
227-097-4
UN:
3439
ADR:
6.1,III
:
163.18 g/mol